Human SELENBP1 (Selenium Binding Protein 1) ELISA Kit
77517-110
:
- Assay Type:Sandwich
- Host:Human
- Target protein:SELENBP1 (Selenium Binding Protein 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway
- Sensitivity:0.055 ng/ml
- Regulatory Status:RUO
- Cat. No.:77517-110