Rat IRF1 (Interferon Regulatory Factor 1) ELISA Kit
Catalog # 77528-568
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:IRF1 (Interferon Regulatory Factor 1)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Apoptosis, tumor immunity, infection immunity
- Sensitivity:0.056 ng/ml
- Regulatory Status:RUO
- Cat. No.:77528-568