Mouse HSP90aA1 (Heat Shock Protein 90kDa Alpha A1) ELISA Kit
Catalog # 77528-388
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:HSP90aA1 (Heat Shock Protein 90kDa Alpha A1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction
- Sensitivity:0.65 ng/ml
- Regulatory Status:RUO
- Cat. No.:77528-388