Human ICD (IsocitRate Dehydrogenase) ELISA Kit
Catalog # 77526-492
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:ICD (IsocitRate Dehydrogenase)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.32 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and kinase
- Sensitivity:0.49 ng/ml
- Regulatory Status:RUO
- Cat. No.:77526-492