Mouse EGFL7 (EGF Like Domain Protein, Multiple 7) ELISA Kit
Catalog # 77525-148
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:EGFL7 (EGF Like Domain Protein, Multiple 7)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction
- Sensitivity:0.062 ng/ml
- Regulatory Status:RUO
- Cat. No.:77525-148