Pig TTR (Transthyretin) ELISA Kit
Catalog # 77522-400
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Pig
- Target protein:TTR (Transthyretin)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:1.57 - 100 ng/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and kinase, tumor immunity, infection immunity, hormone metabolism
- Sensitivity:0.71 ng/ml
- Regulatory Status:RUO
- Cat. No.:77522-400