Human APOL2 (Apolipoprotein L2) ELISA Kit
Catalog # 77519-112
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:APOL2 (Apolipoprotein L2)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway
- Sensitivity:0.067 ng/ml
- Regulatory Status:RUO
- Cat. No.:77519-112