Mouse AGRP (Agouti Related Protein) ELISA Kit
Catalog # 77516-940
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:AGRP (Agouti Related Protein)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, metabolic pathway, neuro science, hormone metabolism
- Sensitivity:11.7 pg/ml
- Regulatory Status:RUO
- Cat. No.:77516-940