Human AGTR1 (Angiotensin II Receptor 1) ELISA Kit
Catalog # 77513-676
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:AGTR1 (Angiotensin II Receptor 1)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.16 - 10 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, metabolic pathway, endocrinology, cardiovascular biology, hormone metabolism
- Sensitivity:0.055 ng/ml
- Regulatory Status:RUO
- Cat. No.:77513-676