Simian TNFa (Tumor Necrosis Factor Alpha) ELISA Kit
Catalog # 77511-660
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Monkey
- Target protein:TNFa (Tumor Necrosis Factor Alpha)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:7.82 - 500 pg/ml
- Storage Temperature:2…8 °C
- Application:Cytokine, tumor immunity, infection immunity
- Sensitivity:2.8 pg/ml
- Regulatory Status:RUO
- Cat. No.:77511-660
Specifications
About this item
- Host: Simian (monkey)