Rat cAMP (Cyclic Adenosine Monophosphate) ELISA Kit
Catalog # 77510-952
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Competitive
- Host:Rat
- Target protein:cAMP (Cyclic Adenosine Monophosphate)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:2.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:312.5 - 20000 pg/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction, metabolic pathway
- Sensitivity:143.2 pg/ml
- Regulatory Status:RUO
- Cat. No.:77510-952