Lipoteichoic Acids (LTA) ELISA Kit
Catalog # 77526-718
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Competitive
- Target protein:Lipoteichoic Acids (LTA)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.31 - 20 ng/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and kinase
- Sensitivity:0.13 ng/ml
- Regulatory Status:RUO
- Cat. No.:77526-718
Specifications
About this item
- Host: General