Mouse PGE2 (Prostaglandin E2) ELISA Kit
Catalog # 77526-538
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Competitive
- Host:Mouse
- Target protein:PGE2 (Prostaglandin E2)
- Size:96 wells
- Sample Type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:3.13 - 200 pg/ml
- Storage Temperature:2…8 °C
- Application:Metabolic pathway, infection immunity, endocrinology, hormone metabolism
- Sensitivity:0.97 pg/ml
- Regulatory Status:RUO
- Cat. No.:77526-538