Rat CTSS (Cathepsin S) ELISA Kit
Catalog # 77523-502
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:CTSS (Cathepsin S)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:62.5 - 4000 pg/ml
- Storage Temperature:2…8 °C
- Application:Enzyme and kinase, tumor immunity, infection immunity, immune molecule
- Sensitivity:23.9 pg/ml
- Regulatory Status:RUO
- Cat. No.:77523-502