Order Entry
United States
ContactUsLinkComponent
Mouse PRKAa1 (Protein Kinase, AMP Activated Alpha 1) ELISA Kit
Mouse PRKAa1 (Protein Kinase, AMP Activated Alpha 1) ELISA Kit
Catalog # 77519-602
Supplier:  AFG Bioscience
CAS Number:  
undefined
Mouse PRKAa1 (Protein Kinase, AMP Activated Alpha 1) ELISA Kit
Catalog # 77519-602
Supplier:  AFG Bioscience
Supplier Number:  EK244593
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Mouse
  • Target protein:
    PRKAa1 (Protein Kinase, AMP Activated Alpha 1)
  • Size:
    96 wells
  • Sample Type:
    Tissue homogenates, cell lysates and other biological fluids
  • Time to Results:
    3.5 h
  • Shelf Life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection Range:
    1.57 - 100 ng/ml
  • Storage Temperature:
    2…8 °C
  • Application:
    Enzyme and kinase, metabolic pathway
  • Sensitivity:
    0.67 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77519-602

Specifications

About this item