Human CRISP3 (Cysteine Rich Secretory Protein 3) ELISA Kit
Catalog # 77517-370
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:CRISP3 (Cysteine Rich Secretory Protein 3)
- Size:96 wells
- Sample Type:Serum, plasma, saliva, sweat and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.94 - 60 ng/ml
- Storage Temperature:2…8 °C
- Application:Signal transduction
- Sensitivity:0.43 ng/ml
- Regulatory Status:RUO
- Cat. No.:77517-370