Human S100A7 (S100 Calcium Binding Protein A7) ELISA Kit
Catalog # 77515-414
Supplier: AFG Bioscience
New Product
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Sandwich
- Host:Human
- Primary antibody reactivity:
- Target protein:S100A7 (S100 Calcium Binding Protein A7)
- Size:96 wells
- Environmentally Preferable:
- Sample Type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:125 - 8000 pg/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, infection immunity
- Sensitivity:50 pg/ml
- Regulatory Status:RUO
- Cat. No.:77515-414