Mouse MUC2 (Mucin 2) ELISA Kit
Catalog # 77513-830
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:MUC2 (Mucin 2)
- Size:96 wells
- Sample Type:Tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.79 - 50 ng/ml
- Storage Temperature:2…8 °C
- Application:Tumor immunity, infection immunity, gastroenterology
- Sensitivity:0.3 ng/ml
- Regulatory Status:RUO
- Cat. No.:77513-830