Mouse VCAM1 (Vascular Cell Adhesion Molecule 1) ELISA Kit
Catalog # 77511-958
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:VCAM1 (Vascular Cell Adhesion Molecule 1)
- Size:96 wells
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf Life:6 months (4 °C); 12 months (–20 °C)
- Detection Range:0.4 - 25 ng/ml
- Storage Temperature:2…8 °C
- Application:CD and adhesion molecule, tumor immunity, infection immunity
- Sensitivity:0.14 ng/ml
- Regulatory Status:RUO
- Cat. No.:77511-958