Cetrimide Selective/MacConkey/Vogel and Johnson Agar triplate
89407-452
:
- Pack type:Triplate
- Description:7 ml/section
- Storage Conditions:2...8 °C
- Cat. No.:89407-452
- Plate size:1.5×10 cm
- Supplier no.:J314
For isolation and differential of gram-negative bacteria.
Section III: Vogel and Johnson Agar; For selective isolation of coagulase-positive and mannitol-positive S. aureus.