Mag-Bind® FFPE DNA Kit
Supplier: Omega Bio-Tek
Certificates
About this item
The Mag-Bind® FFPE DNA 96 kit is designed for rapid and reliable isolation of DNA from formalin-fixed, paraffin-embedded (FFPE) tissue.
- Release short and long DNA fragments
- Reverse crosslinking
- Automation-friendly extraction from FFPE tissue in under six hours
The Mag-Bind® FFPE DNA 96 Kit is designed for rapid and reliable isolation of DNA from formalin-fixed, paraffin-embedded (FFPE) tissue.
This system is fully compatible on Hamilton Star, Beckman Coulter Biomek® FX, Biomek® 2000, Tecan Freedom Evo®, Thermo KingFisher® Flex and other workstations.
The specially formulated buffers reverse crosslinking and release short and long DNA fragments. Mag-Bind® Particles allow for automation-friendly extraction from FFPE tissue in under six hours. Purified DNA is suitable for downstream applications including SNP analysis, sequencing, and genotyping.
Frequently Bought Together