Order Entry
United States
ContactUsLinkComponent
KingFisher™ mL Magnetic Particle Processor
KingFisher™ mL Magnetic Particle Processor
Catalog # 14230-362
undefined
KingFisher™ mL Magnetic Particle Processor
Catalog # 14230-362
Supplier Number:  5400050

Specifications

  • Description:
    KingFisher™ mL magnetic particle processor
  • Electrical:
    110 - 240 V
  • W×D×H:
    30×30×30 cm (11¹³/₁₆×11¹³/₁₆×11¹³/₁₆")
  • Weight:
    11.5 kg (25.4 lbs.)
  • Cat. No.:
    14230-362
  • Supplier no.:
    5400050

Specifications

About this item

Purifies DNA, RNA, proteins, or cells in strips of five tubes.

Processor uses the principle of shifting magnetic particles through purification phases and produces no cross-contamination or reagent carryover. Ideally suited to high-volume samples of 50 µl to 1 ml, with up to 15 samples per run.

The volume range of 50 to 1000 µl per tube enables cells and proteins from larger starting volumes to be isolated with simultaneous concentration. Also ideal for clinical research laboratories that isolate larger amounts of genomic DNA with magnetic particle methods.

Features 3×5 magnetic rods, ready-made protocols, and ability to operate on special 1×5 format tubes. Processor includes 3×5 separate disposable tube strips of five tubes per strip and disposable tip combs with 3×5 format.