Orders LinkContactUsLinkComponent
Cubis® II Semi-Micro Balances
Cubis® II Semi-Micro Balances
Supplier:   Sartorius
Cubis® II Semi-Micro Balances
Supplier:  Sartorius
Certificates
Cubis® II Semi-Micro Balances
36 Options Available Below
Product Details & Documents
Semi-micro balances come with a readability of 0.01 mg or 10 µg and a maximum weighing capacity of up to 220 g.
  • Superior weighing performance: Fast measurement time and accurate weighing results are guaranteed thanks to next generation monolithic weighing system and the engineering solution compensating for environmental changes
  • No more issues with charged samples: Glass draft shields coated with conductive layer prevent outer electrostatic effects and the built-in ionizer effectively eliminates electrostatic charges from samples
  • Cleaning process guidance: Intuitive process for daily and advanced cleaning is now available in all Cubis® II MCA. The cleaning QApp provides visual guidance, information about chemical compatibility as well as electronical tracking of these events
  • Hardware upgradeability: Hardware features like motorized draft shield function or built-in ionizer can be activated post-purchase. An automated inner draft shield is available as a klick-in accessory which is recommended for use under challenging environmental conditions (e.g. in drafty environments)
  • Compliance and Data Integrity: The end-to-end data integrity, technical controls for 21 CFR Part 11 compliance, integrated audit-trail and state-of-the-art user management ensure laboratories can meet regulatory requirements. These features are directly available on the Cubis® II balances, without the need of additional software
  • Balances Fleet Management: The Ingenix Suite is a flexible, open solution that works with or without an ELN/LIM system. It offers unlimited connections to easily manage the entire Cubis® II MCA lab balance fleet across all labs within the same network
  • Ease of use: The teaching function with learning capability of the motorized draft shield, guided workflows for many different applications (QApps), automated motorized leveling and automatic internal adjustment (isoCAL) help with easy and error-free operation of the balance
Semi-micro balances are used for weighing small sample amounts down to the µg-range. To meet the requirements of your specific application, specially designed weighing pans, sample holders and software applications are available to simplify your weighing processes and laboratory workflows.
Specifications for Product Family
Specifications
  • Catalog No:
  • 77476-488
  • 77476-490
  • 77476-492
  • 77476-494
  • 77476-540
  • 77476-542
  • 77476-544
  • 77476-496
  • 77476-498
  • 77476-500
  • 77476-502
  • 77476-504
  • 77476-506
  • 77476-508
  • 77476-510
  • 77476-546
  • 77476-548
  • 77476-550
  • 77476-552
  • 77476-512
  • 77476-514
  • 77476-516
  • 77476-518
  • 77476-520
  • 77476-522
  • 77476-524
  • 77476-526
  • 77476-528
  • 77476-530
  • 77476-532
  • 77476-534
  • 77476-536
  • 77476-538
  • 77476-554
  • 77476-556
  • 77476-558
  • Operating Temperature Range:
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • Plug Type:
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • IEC plug
  • Communication Interface:
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • USB-A;USB-B;USB-C Interface
  • Voltage:
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
Specifications
Product Family Options
Draft shield D
Product Information
  • Weighing capacity
    Linearity
    Pan/Platform Size
    Readability
    Stabilization time
    Repeatabilty
    Dimensions
    Availability
    Price
  • 77476-488
    Semi-micro balance
    Semi-micro balance
    120 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $19,754.65
    $19,754.65
    Each
       $19,754.65
    Minimum order quantity is 1
    Supplier Number: MCA125S3S00D
    Specifications:
    More
    Specifications
    Cat. No.
    77476-488
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-490
    Semi-micro balance
    Semi-micro balance
    120 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $20,881.97
    $20,881.97
    Each
       $20,881.97
    Minimum order quantity is 1
    Supplier Number: MCA125S3S00DHWL
    Specifications:
    More
    Specifications
    Cat. No.
    77476-490
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-492
    Semi-micro balance
    Semi-micro balance
    120 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $20,265.17
    $20,265.17
    Each
       $20,265.17
    Minimum order quantity is 1
    Supplier Number: MCA125S3S00DION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-492
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-494
    Semi-micro balance
    Semi-micro balance
    120 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $20,521.62
    $20,521.62
    Each
       $20,521.62
    Minimum order quantity is 1
    Supplier Number: MCA125S3S00DMDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-494
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-540
    Semi-micro balance
    Semi-micro balance
    120 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $18,645.10
    $18,645.10
    Each
       $18,645.10
    Minimum order quantity is 1
    Supplier Number: MCE125S3S00D
    Specifications:
    More
    Specifications
    Cat. No.
    77476-540
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-542
    Semi-micro balance
    Semi-micro balance
    120 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $19,155.63
    $19,155.63
    Each
       $19,155.63
    Minimum order quantity is 1
    Supplier Number: MCE125S3S00DION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-542
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-544
    Semi-micro balance
    Semi-micro balance
    120 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $19,412.07
    $19,412.07
    Each
       $19,412.07
    Minimum order quantity is 1
    Supplier Number: MCE125S3S00DMDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-544
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-496
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $21,198.09
    $21,198.09
    Each
       $21,198.09
    Minimum order quantity is 1
    Supplier Number: MCA225P3S00D
    Specifications:
    More
    Specifications
    Cat. No.
    77476-496
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-498
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $22,325.42
    $22,325.42
    Each
       $22,325.42
    Minimum order quantity is 1
    Supplier Number: MCA225P3S00DHWL
    Specifications:
    More
    Specifications
    Cat. No.
    77476-498
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-500
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $21,708.63
    $21,708.63
    Each
       $21,708.63
    Minimum order quantity is 1
    Supplier Number: MCA225P3S00DION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-500
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-502
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $21,965.08
    $21,965.08
    Each
       $21,965.08
    Minimum order quantity is 1
    Supplier Number: MCA225P3S00DMDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-502
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-504
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $24,430.96
    $24,430.96
    Each
       $24,430.96
    Minimum order quantity is 1
    Supplier Number: MCA225P3S00DQP99
    Specifications:
    More
    Specifications
    Cat. No.
    77476-504
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-506
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $25,558.27
    $25,558.27
    Each
       $25,558.27
    Minimum order quantity is 1
    Supplier Number: MCA225P3S00DQP99HWL
    Specifications:
    More
    Specifications
    Cat. No.
    77476-506
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-508
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $24,941.48
    $24,941.48
    Each
       $24,941.48
    Minimum order quantity is 1
    Supplier Number: MCA225P3S00DQP99ION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-508
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-510
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $25,197.93
    $25,197.93
    Each
       $25,197.93
    Minimum order quantity is 1
    Supplier Number: MCA225P3S00DQP99MDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-510
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-546
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $20,088.54
    $20,088.54
    Each
       $20,088.54
    Minimum order quantity is 1
    Supplier Number: MCE225P3S00D
    Specifications:
    More
    Specifications
    Cat. No.
    77476-546
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-548
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $21,215.87
    $21,215.87
    Each
       $21,215.87
    Minimum order quantity is 1
    Supplier Number: MCE225P3S00DHWL
    Specifications:
    More
    Specifications
    Cat. No.
    77476-548
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-550
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $20,599.08
    $20,599.08
    Each
       $20,599.08
    Minimum order quantity is 1
    Supplier Number: MCE225P3S00DION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-550
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-552
    Semi-micro balance
    Semi-micro balance
    220|120 g
    0.03 mg
    Ø 90 mm
    0.1|0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $20,855.53
    $20,855.53
    Each
       $20,855.53
    Minimum order quantity is 1
    Supplier Number: MCE225P3S00DMDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-552
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-512
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $26,888.40
    $26,888.40
    Each
       $26,888.40
    Minimum order quantity is 1
    Supplier Number: MCA225S3S00D
    Specifications:
    More
    Specifications
    Cat. No.
    77476-512
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-514
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $28,015.71
    $28,015.71
    Each
       $28,015.71
    Minimum order quantity is 1
    Supplier Number: MCA225S3S00DHWL
    Specifications:
    More
    Specifications
    Cat. No.
    77476-514
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-516
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $27,398.93
    $27,398.93
    Each
       $27,398.93
    Minimum order quantity is 1
    Supplier Number: MCA225S3S00DION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-516
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-518
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $27,655.38
    $27,655.38
    Each
       $27,655.38
    Minimum order quantity is 1
    Supplier Number: MCA225S3S00DMDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-518
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-520
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $31,251.25
    $31,251.25
    Each
       $31,251.25
    Minimum order quantity is 1
    Supplier Number: MCA225S3S00DQP99
    Specifications:
    More
    Specifications
    Cat. No.
    77476-520
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-522
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $30,631.77
    $30,631.77
    Each
       $30,631.77
    Minimum order quantity is 1
    Supplier Number: MCA225S3S00DQP99ION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-522
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-524
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $30,888.23
    $30,888.23
    Each
       $30,888.23
    Minimum order quantity is 1
    Supplier Number: MCA225S3S00DQP99MDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-524
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-526
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 50 mm
    0.005 mg
    1.5 s
    0.004 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $31,373.84
    $31,373.84
    Each
       $31,373.84
    Minimum order quantity is 1
    Supplier Number: MCA226S3S00D
    Specifications:
    More
    Specifications
    Cat. No.
    77476-526
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-528
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 50 mm
    0.005 mg
    1.5 s
    0.004 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $32,501.15
    $32,501.15
    Each
       $32,501.15
    Minimum order quantity is 1
    Supplier Number: MCA226S3S00DHWL
    Specifications:
    More
    Specifications
    Cat. No.
    77476-528
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-530
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 50 mm
    0.005 mg
    1.5 s
    0.004 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $31,884.36
    $31,884.36
    Each
       $31,884.36
    Minimum order quantity is 1
    Supplier Number: MCA226S3S00DION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-530
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-532
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 50 mm
    0.005 mg
    1.5 s
    0.004 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $32,140.81
    $32,140.81
    Each
       $32,140.81
    Minimum order quantity is 1
    Supplier Number: MCA226S3S00DMDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-532
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-534
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 50 mm
    0.005 mg
    1.5 s
    0.004 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $34,606.69
    $34,606.69
    Each
       $34,606.69
    Minimum order quantity is 1
    Supplier Number: MCA226S3S00DQP99
    Specifications:
    More
    Specifications
    Cat. No.
    77476-534
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-536
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 50 mm
    0.005 mg
    1.5 s
    0.004 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $35,368.32
    $35,368.32
    Each
       $35,368.32
    Minimum order quantity is 1
    Supplier Number: MCA226S3S00DQP99ION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-536
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-538
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 50 mm
    0.005 mg
    1.5 s
    0.004 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $35,373.67
    $35,373.67
    Each
       $35,373.67
    Minimum order quantity is 1
    Supplier Number: MCA226S3S00DQP99MDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-538
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-554
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $25,778.85
    $25,778.85
    Each
       $25,778.85
    Minimum order quantity is 1
    Supplier Number: MCE225S3S00D
    Specifications:
    More
    Specifications
    Cat. No.
    77476-554
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-556
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $26,289.38
    $26,289.38
    Each
       $26,289.38
    Minimum order quantity is 1
    Supplier Number: MCE225S3S00DION
    Specifications:
    More
    Specifications
    Cat. No.
    77476-556
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
  • 77476-558
    Semi-micro balance
    Semi-micro balance
    220 g
    0.03 mg
    Ø 90 mm
    0.01 mg
    1.5 s
    0.007 mg
    24×30.1×30.1 cm
    Log in for availability
    Log in for availability
    Each
    Each - $26,545.83
    $26,545.83
    Each
       $26,545.83
    Minimum order quantity is 1
    Supplier Number: MCE225S3S00DMDS
    Specifications:
    More
    Specifications
    Cat. No.
    77476-558
    Operating Temperature Range
    10…30 °C
    Plug Type
    IEC plug
    Communication Interface
    USB-A;USB-B;USB-C Interface
    Voltage
    100 - 240 V
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Pipette calibration kit for ultra-high resolution balances
    Pipette calibration kit for ultra-high resolution balances
    77476-562
    Log in for availability
    Each - $2,226.26
Product Information
  • Catalog #
    Availability
    Price
  • Pipette cleaning kit for ultra-high resolution balances
    Pipette cleaning kit for ultra-high resolution balances
    77476-560
    Log in for availability
    Each - $63.40
Product Information
  • Catalog #
    Availability
    Price
  • Motion sensor
    Motion sensor
    76413-008
    Log in for availability
    Each - $574.01