ContactUsLinkComponent
Engino® STEM Solar Power Complete Set
Engino® STEM Solar Power Complete Set
  470356-018
undefined
Engino® STEM Solar Power Complete Set
  470356-018
 :  UNE951
  • Description:
    Engino STEM solar power complete set
  • Cat. No.:
    470356-018
Use the free energy from the sun to power your designs.
  • Download easy-to-follow instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Climate change and global warming urge scientists to take action and save our planet by reducing carbon emissions. The sun can provide free energy for all of our needs, however, we need to convert it to a useful form.
This set includes one 3 V solar panel that powers an electrical motor, along with digital instructions for 16 solar powered models such as a chopper car, a fan, a plane, a robot and a winch crane. With easy to-follow digital building instructions and detailed explanations of the different scientific principles applied, along with innovative experimental activities, students learn by doing all about Solar Energy while acquiring 21st century skills.
Suggested for students working in groups of 2 or 3.
 : Kit includes 142 plastic parts, 627 connecting points, one DC motor with option to operate in-house with batteries, one 3 V solar panel with cable, digital instructions for 16 models.