Ward's Stream Table Bundle
This unique kit includes a stream table and a starter pack of sediments, with refill options available.
- Specifically designed to work with the NYS earth and space science investigations
- Kit includes a starter pack of soil, fine sand, coarse sand, activity guide, and soil types poster
- 15 pounds of sediment in a bucket with a scoop available as a refill options
- Stream table designed for 2 to 4 students at a time
This stream table kit is outstanding for demonstrating stream morphology, erosion, and deposition. This 14×46×2.5" stream table includes plastic tray, drain trough, tray support, an integrated prop, and siphon tube. Students will learn the impact water has on the development of land formations.
This kit allows students to create their own landforms while simulating water streams and investigating the impact of water on different substrates.
The soil types poster is an excellent supplement to the investigations and visual guide to Earth's soil composition.
: Stream table kit includes: A stream table, 200 g of bagged soil, 32 oz. container of fine sand (two pounds), 32 oz. container of coarse sand (two pounds), 32 oz. container of pebbles (2 pounds), an activity set (complete with a teacher's guide and student copymasters), and an 11×17" Soil Types poster.
Refill buckets are available for each type of sediment. Each bucket contains 15 pounds of material and a scoop. Bundle and save by purchasing all five Refill Buckets together.
This unique kit includes a stream table and a starter pack of sediments, with refill options available.
- Specifically designed to work with the NYS earth and space science investigations
- Kit includes a starter pack of soil, fine sand, coarse sand, activity guide, and soil types poster
- 15 pounds of sediment in a bucket with a scoop available as a refill options
- Stream table designed for 2 to 4 students at a time
This stream table kit is outstanding for demonstrating stream morphology, erosion, and deposition. This 14×46×2.5" stream table includes plastic tray, drain trough, tray support, an integrated prop, and siphon tube. Students will learn the impact water has on the development of land formations.
This kit allows students to create their own landforms while simulating water streams and investigating the impact of water on different substrates.
The soil types poster is an excellent supplement to the investigations and visual guide to Earth's soil composition.
: Stream table kit includes: A stream table, 200 g of bagged soil, 32 oz. container of fine sand (two pounds), 32 oz. container of coarse sand (two pounds), 32 oz. container of pebbles (2 pounds), an activity set (complete with a teacher's guide and student copymasters), and an 11×17" Soil Types poster.
Refill buckets are available for each type of sediment. Each bucket contains 15 pounds of material and a scoop. Bundle and save by purchasing all five Refill Buckets together.
Product Family Options
- Description
Recirculating pump
- Description
- : 6600-01
Refill buckets
- Description
- Coarse sand, 15 lbs.Each (15lb)$39.99Pallet of 80 (15lb)$3,039.19
- Fine sand, 15 lbs.Each (15lb)$39.99Pallet of 80 (15lb)$3,039.19
- Pebbles, 15 lbs.Each (15lb)$39.99Pallet of 80 (15lb)$3,039.19
- Potting soil, 15 lbs.Each (15lb)$49.99Pallet of 36 (15lb)$1,139.79
- Aquarium gravel, 15 lbs.Each (15lb)$39.99Pallet of 80 (15lb)$3,199.19
- Refill bucket bundleEach$159.99Pallet of 264$37,999.99