ContactUsLinkComponent
Buck Hammer
  470334-888
undefined
Buck Hammer
  470334-888
 :  35-133
  • Description:
    Buck hammer
  • Cat. No.:
    470334-888
A standard for reflex testing.
  • Stainless steel handle
  • Incorporated pin and brush testing
For testing reflex response and neurological sensitivity. This two-sided hammer has a large and a small rounded head.
In the handle is a brush for testing touch sensitivity. On the head of the hammer is a needle for testing pain sensitivity. Note the needle included with this hammer is not intended to break the skin.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.