Perfect for venipuncture, I.V. starts, renal dialysis, pre-op prep, and other minor invasive procedures, these pads and swabsticks provide better patient comfort.
- Non-Irritating
- for Minor Invasive Procedures
- PVP Iodine Provides Long Germicidal Activity
- Scrub Swabsticks Improve Prep Pads and Swabsticks' Efficiency
- Prep Pads and Swabsticks Saturated with a 10% Povidone-Iodine Prep Solution
PVP iodine provides longer germicidal activity than ordinary iodine solutions
Non-irritating and virtually non-stinging prep pads and prep swabsticks are saturated with a 10% povidone-iodine prep solution (equivalent to 1% available iodine) to provide proven antisepsis. Scrub swabsticks are saturated with a 7.5% povidone-iodine scrub solution with detergents (equivalent to 0.75% available iodine).
Scrub swabsticks scour away dead skin, dirt, and oil, so prep pads and swabsticks can be more effective.