ContactUsLinkComponent
VWR® Wash Bottle Carrier
VWR® Wash Bottle Carrier
Catalog # 16649-980
Supplier:  VWR International
undefined
VWR® Wash Bottle Carrier
Catalog # 16649-980
Supplier:  VWR International
Specifications
  • Description:
    PP wash bottle carrier for 6 bottles
  • Cat. No.:
    16649-980
  • L×W×H:
    28.6×19.4×22.5 cm (11½×7⅝×8⅞")
Specifications
About this item
Sturdy, lightweight polypropylene carrier can hold up to six wash bottles.
  • Polypropylene construction
  • Sturdy
  • Lightweight
  • Secure, comfortable handles
Its secure handle provides comfort and control. Carrier is chemical-resistant. It holds both 237 mL (8 oz.) and 473 mL (16 oz.) bottles. Autoclavable at 121°C (250°F).