Specifications
- L×W:7.5 × 15"
- Environmentally Preferable:
- Cat. No.:15648-998
- Packaging:65 XL wipes per canister; 6 canisters per case
- Supplier no.:Q85484
Specifications
About this item
These EPA-registered, alcohol-free wipes are effective against over 100 microorganisms, including the human Hepatitis B virus.
- Perfect wipe size for larger surface areas
- Multi-count canister accommodates frequent usage
Ideal for use on hard, nonporous environmental surfaces found in healthcare settings.
Compliant with CDC and OSHA guidelines.