Order Entry
United States
ContactUsLinkComponent
VersaBrite™ Swivel Head Pocket Light, Pelican™
VersaBrite™ Swivel Head Pocket Light, Pelican™
Catalog # 66005-560
Supplier:  Avantor
CSA Certified
undefined
VersaBrite™ Swivel Head Pocket Light, Pelican™
Catalog # 66005-560
Supplier:  Avantor
CSA Certified

Specifications

  • Description:
    Head pocket light, 7 lumen
  • Length:
    9.8 cm (3⅞")
  • Environmentally Preferable:
  • Cat. No.:
    66005-560
  • Weight:
    90.7 g (3.2 oz.) with batteries
  • Supplier no.:
    2250-010-110

Specifications

About this item

Consists of a multiple position, O-ring sealed swivel head and an impact- and water-resistant Xenoy®/polycarbonate case

Note: Not for diving applications.

Features a xenon lamp module, multiple use cap/clothing clip, BugsAway™ red lens, and a Velcro® mounting system. Color: non-glare black.

Certifications: FM and MSHA approved non-incendive. CSA certified.
Ordering information: Includes an adjustable hardhat headband, a removable saddle magnet, and two AA size batteries.