About this item
This container with hinged lid can accommodate larger sizes of chemotherapy waste and sharps
Meets or exceeds JCAHO, OSHA, and NIOSH/CDC standards and requirements.
The supplied sealing gasket and absorbent pad help contain excess liquid and guard against spills. Container features a chemotherapy waste label. Yellow container with white lid.