Multi-Use Nestable Sharps Collectors, BD Medical
Supplier: BD
Certificates
About this item
Units feature a tapered neck and unique one-way safety valve opening, designed to minimize needle pop-back and reduce overfilling
Nestable design reduces storage space requirements. Constructed of puncture-resistant polypropylene with clear tops, the units are completely disposable by autoclaving, compaction, or incineration according to institutional procedures.
For accessories, see BD305451 series.
Recessed flanges, a lockable safety enclosure, and sturdy handles make it unnecessary for hands to come into contact with the body of the collector, reducing the risk of needlestick injury.
Frequently Bought Together