ContactUsLinkComponent
Presaturated Wiper, CleanTex™
Presaturated Wiper, CleanTex™
Catalog # TWTX9401
Supplier:  Advantus
undefined
Presaturated Wiper, CleanTex™
Catalog # TWTX9401
Supplier:  Advantus
Supplier Number:  CT9401
Specifications
  • Cat. No.:
    TWTX9401
  • Packaging:
    100/FlexPack
  • W×L:
    22.9 x 27.9 cm (9 x 11")
  • Supplier no.:
    CT9401
Specifications
About this item
A strong, polyester-blend wiper developed for general surface cleaning applications.
  • Presaturated with a 70/30 blend of isopropyl alcohol and water
Wipers are packaged in convenient dispenser packages of 100 wipers each.