Plastibox® Conductive Bins, LEWISBins+™
Supplier: Lewisbins+
Certificates
About this item
ESD-Safe Part Bins are permanently molded in conductive material for use in cleanrooms and workstations.
- Different sizes available
- Protect parts from the damaging effects of static electricity
- Label insert area for easy identification
- Smooth gravity flow interior optimizes parts accessibility
- Bins hang on most louvered panels or rails
Open, sloping hopper fronts for easy product removal. These stackable bins also feature rear hanging lips for attachment to most louvered panels or rails.
Combine Part Bins with ESD-Safe Metal Storage Systems to meet your work-in-process requirements.
Color: Black
Ordering information: Minimum order of 24.
Caution: Note: PB50 is not designed for use on hanging systems.
Frequently Bought Together