Conductive Divider Boxes, LEWISBins+™, Orbis
Permanently conductive containers are made of a strong and durable carbon-filled, injection-molded polypropylene copolymer resin
Ideal for use in commercial, industrial, electronics, and research facilities for storage, assembly, fabrication, and distribution applications. Compatible with many types of racking, shelving, and material handling equipment. Boxes feature a strong stacking ridge and ribbed walls. Two cardholder snaps are standard on each box. Color: black.
: Dividers and covers are also available, contact your Avantor representative for more information.
Frequently Bought Together