Order Entry
United States
ContactUsLinkComponent
Assure™ 425™ Surface-Curing Threadlocker, Loctite®, Henkel
Assure™ 425™ Surface-Curing Threadlocker, Loctite®, Henkel
Catalog # 82002-808
Supplier:  Loctite
undefined
Assure™ 425™ Surface-Curing Threadlocker, Loctite®, Henkel
Catalog # 82002-808
Supplier:  Loctite
Supplier Number:  135461

Specifications

  • Description:
    20 g (0.7 oz.) bottle
  • Viscosity:
    60cP
  • Temperature Range:
    –53...82 °C (–65...180 °F)
  • Cat. No.:
    82002-808
  • Supplier no.:
    42540

Specifications

About this item

This low-strength, fast-curing threadlocker was developed for locking fasteners on plastic or metal parts.

  • Low-strength
  • Fast-curing
  • Can be post-applied

Recommended for nylon, polyester, and other plastic surfaces. Store refrigerated.