Order Entry
United States
ContactUsLinkComponent
Hypersil GOLD™ Peptide HPLC Columns
Hypersil GOLD™ Peptide HPLC Columns
Supplier:  Thermo Scientific
Certificates
Hypersil GOLD™ Peptide HPLC Columns
Supplier:  Thermo Scientific

Specifications

  • Catalog No:
  • TS26002-102130
  • TS26002-152130
  • TS26002-052130
  • Carbon Load:
  • 11%
  • 11%
  • 11%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Max. Pressure:
  • 21756 psi (1500 bar)
  • 21756 psi (1500 bar)
  • 14503 psi (1000 bar)
  • pH Range:
  • 2 - 11
  • 2 - 11
  • 2 - 11
  • Surface Area:
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • Temperature Limit:
  • 60 °C
  • 60 °C
  • 60 °C
  • USP Code:
  • L1
  • L1
  • L1

Specifications

Related Information

Frequently Bought Together