ContactUsLinkComponent
Tempo-iKupffer™ Human iPSC-derived Kupffer Cells
Catalog # 77461-308
CAS Number:  
Tempo-iKupffer™ Human iPSC-derived Kupffer Cells
Catalog # 77461-308
Supplier Number:  1020
CAS Number:  
Specifications
  • Description:
    Human iPSC-derived kupffer cells
  • Cat. No.:
    77461-308
Specifications
About this item
Kupffer cells are the resident macrophages in the human liver. Like Tempo-iPhago™ (macrophages are white blood cells that capture and kill microorganisms), Kupffer cells display innate immune functions protecting the liver from bacterial infections and serve as gatekeepers.
  • Cryovial: ~0.5×10⁶ to ~1.0×10⁶
Tempo-iKupffer™ are reprogrammed from human iPSCs, using integration-free induced pluripotent stem cell (iPSC) lines under a fully defined proprietary serum-free, virus-free, nucleic-acids-free, and integration-free conditions.
They express the following biomarkers using RT/q-PCR and Immunocytochemistry (ICC) methods: CD163, CD206, NR1H3, SpiC, CD68, S100AB.
Intended for scientific research, drug discovery and therapeutics development use only. It is not a product for human testing or diagnostics.
Long-term storage: liquid nitrogen
Growth Properties: 2D/monolayer cell culture technique; 3D/spheroid/organoid cell culture techniques
QC: Sterility, safety (BioSafety Level 2), HIV/viruses, bacteria, fungi: negative. Cell viability post-thawing (>85%)