Tetrahydrofuran, anhydrous ≥99.9%, DriSolv®, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
:Packed in septum-sealed bottle.
Flash Pt:–21.5 °C
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance:Clear liquid, free from particulates
- Assay (GC):99.9% min
- Color (APHA):10 max
- Residue after evaporation:5ppm max
- Expiration date:Exp 12 months from mfg date
- Filtered through 0.2 µm filter:To pass test
- Peroxide (as H2O2) at time of manufacture:0.002% max
- UV Cut-off:212nm max
- Water (H2O) at time of manufacture:0.002% max
Specification Test Results
Related Information
Certificates
Search by Lot NumberDriSolv Accessories Technical Drawing
Technical Drawing
DriSolv Description
Datasheet
EMD Millipore DriSolv Anhydrous Solvents Brochure
Product Literature
ghs_TX0277
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together