1,2-Dimethoxyethane, anhydrous ≥99.5%, DriSolv®, Supelco®
Catalog # EM-DX1531-6
Supplier: Avantor
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:EM-DX1531-6
- Supplier no.:DX1531-6
Specifications
Specification Test Results
- Expiration date:Exp 60 months from mfg date
- Assay (GC):99.5% min
- Appearance:Clear liquid, free from particulates
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Residue after evaporation:5ppm max
- Water (H2O):50ppm max
Specification Test Results
About this item
:
:90.12 g/mol
Merck Index:13,03251
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–69 °C
Density:1.113 g/cm³ (20 °C)
Flash Pt:–6 °C
Storage Temperature:Ambient
EINECS:203-794-9
UN:2252
ADR:3,II
:Packed in septum-sealed bottle.
:GHS02
:GHS08
:GHS07
CAS Number:110-71-4
Formula:C₄H₁₀O₂
MDL Number:MFCD00008502