ContactUsLinkComponent
Oil Sample Bottle, Qorpak
Oil Sample Bottle, Qorpak
  16455-000
 :  Qorpak
Small Business Enterprise
undefined
Oil Sample Bottle, Qorpak
  16455-000
 :  Qorpak
 :  GLA-00856
Small Business Enterprise
  • Capacity:
    120 ml (4 oz.)
  • Cat. No.:
    16455-000
  • Supplier no.:
    GLA-00856
Clear/flint oil sample bottles are provided with a #6 cork and feature a unique design.
  • Oil sample bottle are made of glass
These clear bottles are ideal for field analysis in the petroleum industry. The design of this bottle also makes it an excellent choice for displaying product samples.