Steel Screws and Plastic Wall Anchors for Mounting Steel Pegboard System
Catalog # 77269-716
Supplier: Triton Products
Specifications
- Color:Silver/blue
- Description:Twelve steel screws and twelve plastic wall anchors for mounting steel pegboard system
- Cat. No.:77269-716
Specifications
About this item
Everything needed to mount LocBoard® or steel pegboard to the wall.
- Contains 12 steel screws and 12 plastic wall anchors in one package to mount steel pegboards to any flat wall surface
- Screws are designed to fit through front of square holes to mount the LocBoard tightly to the wall surface
- LocBoard hardware mounting kit is ideal for mounting the stainless steel pegboards on wall studs, hollow wall or concrete block
- Eliminates framing hassles and expense and requires no special tools
LocBoard® mounting hardware kit has everything required to mount LocBoard heavy-duty Epoxy coated steel boards on any flat wall surface. The kit will keep the boards secured to flat wall surfaces. The heads of the screws are sized so that they fit through the front of the square holes and are invisible from the front adding to the LocBoard steel pegboard's clean looking surface.
Ordering information: Limited lifetime warranty.