Stainless Steel Screws and Plastic Wall Anchors for Mounting Stainless Steel Pegboard System
77269-718
- Color:Silver/blue
- Description:Twelve stainless steel screws and twelve plastic wall anchors for mounting stainless steel pegboard system
- Cat. No.:77269-718
Everything needed to mount LocBoard® or steel pegboard to the wall.
- Contains 12 stainless steel screws and 12 plastic wall anchors in 1 package to mount stainless steel pegboards to any flat wall surface
- Stainless steel screws are designed to fit through front of square holes to mount the stainless steel LocBoard tightly to the wall surface
- Stainless Steel LocBoard hardware mounting kit is ideal for mounting the stainless steel pegboards on wall studs, hollow wall or concrete block
- Use with any stainless steel LocBoard® pegboard
Stainless Steel LocBoard® mounting hardware kit has everything required to mount stainless steel heavy-duty pegboard on any flat wall surface. The kit will keep the boards secured to flat wall surfaces. The heads of the stainless steel screws are sized so that they fit through the front square holes of the Stainless Steel LocBoard and are invisible from the front adding to the LocBoard stainless steel pegboard's clean looking surface.
: Limited lifetime warranty.