Nalgene® Large Square Bottle, Polypropylene, Wide Mouth, Thermo Scientific
Catalog # 16130-005
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:4 L (1 gal.)
- Cat. No.:16130-005
- Supplier no.:2122-0010
Specifications
About this item
Space-saving bottle features convenient, molded-in handgrips.
- Excellent for storing and sterilizing contaminated slides and other small lab items
- Not leakproof
- Autoclavable
- Cap size: 100-415
Note: Before autoclaving, set cap on top of the bottle without engaging threads.
Ordering information: Supplied with polypropylene screw caps assembled on bottles.