ContactUsLinkComponent
Xcelite® 99® Handles and Blades, Apex
Xcelite® 99® Handles and Blades, Apex
  37693-878
undefined
Xcelite® 99® Handles and Blades, Apex
  37693-878
 :  997N
  • Description:
    Nutdriver Blade
  • Cat. No.:
    37693-878
  • Blade length:
    92 mm (3⅝")
  • Blade diameter:
    5.6 mm (⁷⁄₃₂")
  • Supplier no.:
    997
With interchangeable blades, shanks, and handles, items can be quickly and easily added or substituted
Regular, tee, and ratchet handles meet every application need. The tee handle enables easy, ergonomic turning action. Ratcheting handles allow simple, quiet, one-handed operation. They also offer smooth, precise action and have a left-right-neutral marking indicator. Light ratcheting drag facilitates turning and prevents fasteners from reversing.