ContactUsLinkComponent
BenchCel Workstation
BenchCel Workstation
Catalog # 77463-590
BenchCel Workstation
Catalog # 77463-590
Supplier Number:  G5590GA
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BenchCel workstation
  • Cat. No.:
    77463-590
Specifications
About this item
The Agilent BenchCel workstation is an automated microplate handler capable of transferring multiple plates at once from a microplate stacker to utilities enabling barcode and other plate labeling, plate centrifugation, and plate sealing. The compact workstation can be used in any lab setting.
  • Two, four, or six labware rack configurations available with the BenchCel microplate handler enhance scalability
  • Platform can be configured with the Agilent PlateLoc thermal microplate sealer, Microplate barcode labeler, or microplate centrifuge
  • Enabled for microplate workflows requiring automated plate sealing, barcode labeling, and plate centrifugation
  • Eight-second transfer time from stack to instrument
  • Adjustable plate capacity handles up to 60 standard SBS microplates per rack
Hands-on time is reduced to a minimum through the automation of routine yet laborious manual microplate management tasks, increasing your lab’s productivity. Save further on setup time with intuitive VWorks software, allowing you to perform microplate stacking and sealing management tasks with confidence and efficiency.