Specifications
- Description:Bravo NGS workstation bundle
- Cat. No.:77463-586
Specifications
About this item
High-throughput (HT) NGS sample preparation robot for increased efficiency and maximized walk away time.
- Quickly automate with ready-to-use protocols and pre-developed methods for Agilent SureSelect, Agilent HaloPlex, Illumina TruSeq, NuGEN Ovation, KAPA HTP, PacBio SMRTbell, NEBNext, and more
- Easy-to-use interface requires no programming expertise to run your automated methods
- Consistent data quality allows you to increase your throughput, with consistent results every time
- Prepare up to ten times more samples in the same time
- Walk-away automation reduces hands-on time by five times compared to manual workflow
- Open platform allows you to automate more NGS or genomics methods with confidence
- The optional on-deck thermal cycler (ODTC) is a compact and configurable accessory on the Bravo NGS system which increases walk-away time and simplifies workflows
The Agilent Bravo NGS Workstation is built on the Bravo automated liquid handling robot preconfigured for library prep and target enrichment using next-generation sequencing (NGS) protocols. Workstation modules add microplate handling to maximize walk away time. Intuitive Agilent VWorks software enables quick setup of preprogrammed protocols yet allows advanced users to create custom protocols.
The Bravo NGS Workstation excels at high-throughput sample prep through the consistency and dependability of the Bravo module. Integrated hardware, like the new on-deck thermal cycler (ODTC), combined with SureSelect protocols streamlines the NGS sample preparation workflow while maintaining application flexibility. Controlled by the same software as the Bravo NGS, your laboratory can seamlessly upgrade to high throughput and maximize the investment in laboratory automation.