Order Entry
United States
ContactUsLinkComponent
 
GemCell™ Plus Xeno-Free Human Serum, Aseptic Assurance System
Catalog # 77463-598
Supplier:  GeminiBio
undefined
GemCell™ Plus Xeno-Free Human Serum, Aseptic Assurance System
Catalog # 77463-598
Supplier:  GeminiBio
Supplier Number:  100-912-750
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    GemCell™ Plus xeno-free, human serum AB - AAS
  • Size:
    200×100 ml
  • Environmentally Preferable:
  • Shipping Conditions:
    Ships on dry ice
  • Origin:
    US
  • Storage Temperature:
    Store frozen at –20...–10 ℃
  • Cat. No.:
    77463-598
  • Packaging:
    BAGA

Specifications

About this item

Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free human serum is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material goes through additional rigorous testing and contains therapeutic grade human thrombin. All donor units are tested for viral markers and found to be non-reactive.

  • This product was developed without the use of animal material
  • Custom fill sizes in bottle and bag format offered
  • 0.1 µm sterile-filtered
  • Supports clinical and commerical applications
  • Aseptic Assurance System