Potassium nitrate, crystals
80700-150
: Avantor
:
- Size:22.7 kg
- Environmentally Preferable:
- Cat. No.:80700-150
- Grade:ACS reagent
- Quality level:200
- Assay:≥99.0%
- Impurities:≤0.005% insolubles
- pH:4.5 - 8.5
: Nitrate of Potash
:
:101.1 g/mol
:7757-79-1
:KNO₃
:MFCD00011409
:334
:13,07733