Order Entry
United States
Orders LinkContactUsLinkComponent
 

Calcium fluoride

22.7 kg
  80700-148
 :   Avantor
Calcium fluoride
  80700-148
 :  Avantor
 :  7789-75-5
Calcium fluoride

22.7 kg

 :  7789-75-5
 
$189.45
   
Each (23kg)
 
  •  :7789-75-5
  •  :CaF₂
  •  :MFCD00010907
  •  :78.07 g/mol
  •  :2500 °C
  •  :3.18 g/ml
 

 

  • Quality level:
    100
  • Grade:
    Certified reference material
  • Format:
    Matric material

 

  • Size:
    22.7 kg